Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TPO Rabbit Polyclonal Antibody

Supplier: BioVision

5351R-100
10007-748EA 252.35 USD InStock
10007-748
Anti-TPO Rabbit Polyclonal Antibody
Antibodies
TPO (Thrombopoietin, also known as Mpl-ligand, MGDF) stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty