Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFR1 Rabbit Polyclonal Antibody

Supplier: BioVision

5348-100
10007-746EA 252.35 USD InStock
10007-746
Anti-TNFR1 Rabbit Polyclonal Antibody
Antibodies
Tumor Necrosis Factor receptor type I (TNF-RI) is also referred as p60. The p60 form of the receptor is primarily expressed on epithelial cells. Recombinant Human soluble TNF receptor Type I (sTNF-RI) is a 18.3 kDa protein containing 162 amino acid residues comprising the extra cellular domain of TNF Receptor Type I.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: TNFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty