Anti-TGFA Rabbit Polyclonal Antibody

Supplier: BioVision
5339-100
10007-732EA 286.04 USD
10007-732
Anti-TGFA Rabbit Polyclonal Antibody
Transforming Growth Factor-alpha (TGF-α) stimulates the proliferation of a wide range of epidermal and epithelial cells. TGF-α has been shown to play a role in the autocrine growth of certain transformed cells.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Neutralization: 2-10 µg/ml

Type: Primary
Antigen: TGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR