Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL5 Rabbit Polyclonal Antibody

Supplier: BioVision

5323-100
10007-722EA 272.95 USD InStock
10007-722
Anti-CCL5 Rabbit Polyclonal Antibody
Antibodies
RANTES (regulated upon activation, normal T-cell expressed and secreted) is a 70-amino acid CC chemokine originally cloned from T-cell-specific sequences-enriched cDNA libraries. Both human and murine RANTES are chemotactic for monocytes. Human RANTES is also a chemoattractant for T-cells and eosinophils, as well as an activator of basophils for histamine release. In contrast to other CC chemokines, RANTES is only slowly expressed after activation of leukocytes. It is involved in a number of pathological conditions like chronic inflammation and autoimmune diseases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CCL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty