Anti-RANKL Rabbit Polyclonal Antibody

Supplier: BioVision
5318R-100
10007-720EA 286.04 USD
10007-720
Anti-RANKL Rabbit Polyclonal Antibody
Human RANK (receptor activator of NF-kB) is a member of the TNFR family, identified as a dentritic cell membrane protein. Human soluble RANK Ligand is a soluble 20 kDa polypeptide, comprising the TNF homologous region of RANKL (176 amino acid residues).

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RANKL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR