Anti-NGN2 Rabbit Polyclonal Antibody

Supplier: BioVision
5307-100
10007-700EA 379.44 USD
10007-700
Anti-NGN2 Rabbit Polyclonal Antibody
Neurogenin-2 (NGN2) is a neural-specific basic helix-loop-helix (bHLH) transcription factor that can specify a neuronal fate on ectodermal cells and is expressed in neural progenitor cells within the developing central and peripheral nervous systems. NGN2 is thoμght to work with Nurr1 to play a role in the differentiation and survival of midbrain dopaminergic neurons. It has also been sμggested for use in human embryonic neural progenitors as a graft for spinal cord injuries

Recommended Dilutions: Western Blot: 1 - 2 ?g/ml, ELISA

Type: Primary
Antigen: NGN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR