Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CITED2 Rabbit Polyclonal Antibody

Supplier: BioVision

5286-100
10007-686EA 334.75 USD InStock
10007-686
Anti-CITED2 Rabbit Polyclonal Antibody
Antibodies
CITED2 was initially identified as a potential transcriptional activator with significant homology to MSG1, a melanocyte-specific nuclear protein. CITED2 interacts with the histone acetyltransferase p300/CBP and acts as a coactivator to several DNA-binding transcription factors such as HIF-1 and AP-2. CITED2 also controls proliferation of mouse embryonic fibroblasts via the polycomb group genes Bmi-1 and Mel18 and the tumor suppressor Ink4a/Arf. CITED2 has also been found to be an essential regulator of adult hematopoietic stem cells (HSCs), with conditional deletion of CITED2 in the adult mouse resulting in the loss of HSCs, multilineage bone marrow failure and increased lethality.

Recommended Dilutions: Western Blot: 1 - 2 μg/ml, ELISA

Type: Primary
Antigen: CITED2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty