Anti-MIP2 Rabbit Polyclonal Antibody

Supplier: BioVision
5253-100
10007-576EA 309.39 USD
10007-576
Anti-MIP2 Rabbit Polyclonal Antibody
MIP-2 (macrophage inflammatory protein-2) is a 69-amino acid CC chemokine originally cloned from LPS-stimulated rat lung. MIP-2 is induced during acute inflammation in rat models of diseases. It is a potent neutrophil chemotactic factor both in vitro and in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml; and neutralization

Type: Primary
Antigen: MIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR