Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MIP2 Rabbit Polyclonal Antibody

Supplier: BioVision

5251-100
10007-574EA 272.95 USD InStock
10007-574
Anti-MIP2 Rabbit Polyclonal Antibody
Antibodies
Murine microphage inflammatory protein-2 (MIP-2) promotes neutrophil chemotaxis and degranulation. Murine MIP-2 is a 7.8 kDa protein containing 73 amino acid residues including the ‘RLR’ motif common to the CXC chemokine family that bind to CXCR1 or CXCR2.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Neutralization: 2-10 µg/ml

Type: Primary
Antigen: MIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty