Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MIP1 beta Rabbit Polyclonal Antibody

Supplier: BioVision

5249-100
10007-572EA 272.95 USD InStock
10007-572
Anti-MIP1 beta Rabbit Polyclonal Antibody
Antibodies
MIP-1β (macrophage inflammatory protein-1β) is a 69-amino acid CC chemokine with a molecular weight of 7.8 kDa. MIP-1β induces migration of monocytes/macrophages, and activates CD4+ cells, but not NK cells and eosinophils.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Immunoprecipitation: 4-8 µg/ml; and neutralization

Type: Primary
Antigen: MIP1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty