Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MIP1 Rabbit Polyclonal Antibody

Supplier: BioVision

5246-100
10007-538EA 272.95 USD InStock
10007-538
Anti-MIP1 Rabbit Polyclonal Antibody
Antibodies
MIP-1α (macrophage inflammatory protein-1α is a 60-amino acid CC chemokine with a molecular weight of 7.9 kDa. MIP-1α induces migration of monocytes/macrophages, B-lymphocytes, activated CD8+ T cells, natural killer (NK) cells, eosinophils. MIP-1α also stimulates basophils to release histamine, induces ICAM-1 expression, mast cell degranulation, and production of TNF-α, IL-1 and IL-6. In addition to its proinflammatory activities, MIP-1α inhibits the proliferation of hematopoietic stem cells in vitro and in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml

Type: Primary
Antigen: MIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty