Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCSF Rabbit Polyclonal Antibody

Supplier: BioVision

5238-100
10007-536EA 252.35 USD InStock
10007-536
Anti-MCSF Rabbit Polyclonal Antibody
Antibodies
Macrophage Colony Stimulating Factor (M-CSF) is produced by various cell types, including fibroblasts, microphages, and endothelial cells. M-CSF is involved in both the development and function of monocytes/microphages. The recombinant M-CSF is a disulfide-linked homodimer composed of two 18.2 kDa subunits of 156 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MCSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty