Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCP1 Rabbit Polyclonal Antibody

Supplier: BioVision

5225-100
10007-532EA 272.95 USD InStock
10007-532
Anti-MCP1 Rabbit Polyclonal Antibody
Antibodies
Rat MCP-1/JE (monocyte chemoattractant protein-1) is a 148-amino acid CC chemokine with a N-terminal sequence of 29 residues as a signal sequence. It was originally cloned from Con-A-stimulated rat spleen cDNA library. This rat MCP-1/JE is 49-amino acid longer than human MCP-1 at 3’-end. The 3’-end is a serine and threonine rich zone, which is probably responsible for the extensive O-glycosylation and which explains for the higher molecular weight (25 kDa). In vitro, MCP-1/JE is chemotactic for monocytes as well as lymphocytes and basophils, but not for neutrophiles. MCP-1/JE is produced by a wide range of cell types as a reaction to diverse inflammatory stimuli.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml;From researcher’s feedback, it can also be used for neutralization studies

Type: Primary
Antigen: MCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty