Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCP1 Rabbit Polyclonal Antibody

Supplier: BioVision

5222-100
10007-528EA 252.35 USD InStock
10007-528
Anti-MCP1 Rabbit Polyclonal Antibody
Antibodies
Human macrophage/monocyte chemotactic and activating factor (MCAF), also called human monocyte chemotactic protein-1 (MCP-1), is an 8.6 kDa protein containing 76 amino acid residues. It plays an important role in the inflammatory response of blood monocytes and tissue microphages.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty