Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIF Rabbit Polyclonal Antibody

Supplier: BioVision

5216-100
10007-526EA 252.35 USD InStock
10007-526
Anti-LIF Rabbit Polyclonal Antibody
Antibodies
Leukemia Inhibitory Factor (LIF) is a lymphoid factor that promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. LIF has several functions such as cholinergic neuron differentiation, control of stem cell pluripotency, bone & fat metabolism, mitogenesis of factor dependent cell lines & promotion of megakaryocyte production in vivo. Human and mouse LIF exhibit a 78% identity in its amino acid sequence. Human LIF is as active on human cells as is it is on mouse cells, thoμgh mouse LIF is about 1000 fold less active on human cells, than human LIF.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; ELISA: 2-3 µg/ml; and Neutralization: 0.5-1 µg/ml

Type: Primary
Antigen: LIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty