Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDH6 Rabbit Polyclonal Antibody

Supplier: BioVision

5211-100
10007-680EA 309 USD InStock
10007-680
Anti-CDH6 Rabbit Polyclonal Antibody
Antibodies
KC, a homolog of human and hamster gro/MGSA, is a 72-amino acid CXC chemokine originally cloned from rat macrophages and lung tissue. It is the mediator for recruitment and activation of netrophils in rat lung inflammation models. Expression of KC can be upregulated by LPS and IL-1β stimulation. IFN-g blocks LPS-induced expression of KC.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 3-5 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: CDH6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty