Anti-CCR10 Rabbit Polyclonal Antibody

Supplier: BioVision
5210-100
10007-678EA 309.39 USD
10007-678
Anti-CCR10 Rabbit Polyclonal Antibody
Recently, a function has been assigned to a previously identified orphan chemokine receptor, GPR-2. This receptor, also called CCR10, binds with monocyte chemoattractant protein (MCP)-1, MCP-3 and MIP-1 beta. CCR10 is expressed by melanocytes, dermal fibroblasts, dermal microvascular endothelial cells, T cells as well as in skin-derived Langerhans cells. CCR10 may play a role in both skin homeostasis as well as a potential role in inflammatory responses.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CCR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR