Anti-IP10 Rabbit Polyclonal Antibody

Supplier: BioVision
5202-100
10007-676EA 309.39 USD
10007-676
Anti-IP10 Rabbit Polyclonal Antibody
IP-10 (interferon-gamma induced protein 10 kDa; in mouse also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml

Type: Primary
Antigen: IP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR