Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IP10 Rabbit Polyclonal Antibody

Supplier: BioVision

5200-100
10007-674EA 319.3 USD InStock
10007-674
Anti-IP10 Rabbit Polyclonal Antibody
Antibodies
Human IP-10 is an 8.5 kDa protein produced by several cell types during the delayed type hypersensitivity response. IP-10 acts as a chemoattractant towards monocytes, lymphocytes, and certain T cells.

Recommended Dilutions: Western Blot: 0.5-1.0 µg/ml; Neutralization: 10 µg/ml; and ELISA: 2-4 µg/ml)

Type: Primary
Antigen: IP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty