Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDNF Rabbit Polyclonal Antibody

Supplier: BioVision

5198-100
10007-672EA 272.95 USD InStock
10007-672
Anti-GDNF Rabbit Polyclonal Antibody
Antibodies
Human GDNF (Glial-Derived Neurotrophic Factor) is a 30.1 kDa protein that specifically promotes dopamine uptake and survival of midbrain neurons. GDNF is a disulphide-linked homodimer, formed by two identical 134 amino acids subunits.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: GDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty