Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL18 Rabbit Polyclonal Antibody

Supplier: BioVision

5180R-100
10007-670EA 338.75 USD InStock
10007-670
Anti-IL18 Rabbit Polyclonal Antibody
Antibodies
IL-18 (IFN-gamma-inducing factor, IGIF) is a novel proinflammatory cytokine that aμgments natural killer (NK) activity in spleen cells. It also has other biological properties such as activation of NF-kB, stimulation of Fas ligand expression, induction of CC and CXC chemokines. IL-18 increases production of competent HIV. IL-18 is related to the IL-1 family. One of the IL-18 receptor chains is the IL-1 receptor-related protein, a member of the IL-1R family. Like IL-1β, IL-18 is first synthesized as a leaderless precursor requiring the IL-1β converting enzyme (ICE) for cleavage into an active molecule. IL-18 may play a role in tissue injury in inflammatory reactions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 2-5 µg/ml; and Neutralization: 2-5 µg/ml

Type: Primary
Antigen: IL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty