Anti-IL17 Rabbit Polyclonal Antibody

Supplier: BioVision
5176-100
10007-662EA 309.39 USD
10007-662
Anti-IL17 Rabbit Polyclonal Antibody
IL-17 was identified from a CD4+ T cells DNA library. IL-17 can be induced from primary peripheral blood CD4+ T cells upon stimulation. The cytokine exhibits a high degree of amino acid identity with HVS13, an open reading frame from a T lymphotropic Herpesvirus saimiri and with murine CTLA8. Supernatant from cells transfected with IL-17 induced IL-16 and IL-18 production and enhanced the surface expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. IL-17 is a disulfide-linked homodimer of two 31.0 kDa subunits, each containing 136 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; ELISA: 2-4 µg/ml

Type: Primary
Antigen: IL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR