Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL15 Rabbit Polyclonal Antibody

Supplier: BioVision

5172-100
10007-660EA 272.95 USD InStock
10007-660
Anti-IL15 Rabbit Polyclonal Antibody
Antibodies
IL-15 is a member of the four alpha-helical bundle family of cytokines with a molecular weight of 14-15 kDa. Its in vitro activity is very similar to IL-2, except that IL-15 utilizes the IL-15 receptor alpha subunit for binding. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes. It also activates a number of important intracellular signaling molecules. This implies that IL-15 could be essential for the immune responses, allograpt rejection, and the pathogenesis of autoimmune diseases.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: IL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty