Anti-IL13 Rabbit Polyclonal Antibody

Supplier: BioVision
5165-100
10007-654EA 309.39 USD
10007-654
Anti-IL13 Rabbit Polyclonal Antibody
Interleukin-13 (IL-13) is an immunoregulatory protein produced by activated T lymphocytes. The biological activities of IL-13 include the stimulation of B cell proliferation and immunoglobulins production.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Neutralization: 2-4 µg/ml

Type: Primary
Antigen: IL13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR