Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL6 Rabbit Polyclonal Antibody

Supplier: BioVision

5145-100
10007-644EA 272.95 USD InStock
10007-644
Anti-IL6 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant rat IL-6. The antibody was purified by affinity chromatography employing immobilized rat IL-6 matrix.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty