Anti-IL6 Rabbit Polyclonal Antibody

Supplier: BioVision
5143-100
10007-640EA 286.04 USD
10007-640
Anti-IL6 Rabbit Polyclonal Antibody
IL-6 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells. Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4.0 µg/ml

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR