Anti-IL4 Rabbit Polyclonal Antibody

Supplier: BioVision
5137-200
10007-638EA 344.42 USD
10007-638
Anti-IL4 Rabbit Polyclonal Antibody
IL-4 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells. Human IL-4 is a 15.0 kDa globular protein containing 130 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR