Anti-IL1B Rabbit Polyclonal Antibody

Supplier: BioVision
5128-100
10007-628EA 297.72 USD
10007-628
Anti-IL1B Rabbit Polyclonal Antibody
IL-1β is a potent immuno-modulator that mediates a wide range of immune and inflammatory responses including the activation of B and T cells. Human IL-1β is a 17.0 kDa protein containing 153 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR