Anti-IL1A Rabbit Polyclonal Antibody

Supplier: BioVision
5125-100
10007-626EA 286.04 USD
10007-626
Anti-IL1A Rabbit Polyclonal Antibody
IL-1α is a potent immuno-modulator that mediates a wide range of immune and inflammatory responses. Human IL-1α is an 18.0 kDa protein containing 159 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml

Type: Primary
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR