Anti-IGF2 Rabbit Polyclonal Antibody

Supplier: BioVision
AB18954-100
10007-624EA 367.24 USD
10007-624
Anti-IGF2 Rabbit Polyclonal Antibody
Antibodies
IGF-II (Insulin-like Growth Factor-II) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types. Human IGF-II is a 7.5 kDa protein containing 67 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR