Anti-IGF1 Rabbit Polyclonal Antibody

Supplier: BioVision
5119-100
10007-620EA 286.04 USD
10007-620
Anti-IGF1 Rabbit Polyclonal Antibody
IGF-I (Insulin-like Growth Factor-I) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types including muscle, bone, and cartilage tissue. Human IGF-I is a 7.6 kDa protein containing 70 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR