Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFNG Rabbit Polyclonal Antibody

Supplier: BioVision

5117-100
10007-618EA 252.35 USD InStock
10007-618
Anti-IFNG Rabbit Polyclonal Antibody
Antibodies
IFN-gamma is an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN-gamma receptors are present in most immune cells, which respond to IFN-gamma signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-gamma signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. Additionally, IFN-gamma stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Murine IFN-gamma is a 15.6 kDa protein containing 134 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Neutralization: 0.2-1 µg/ml

Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty