Anti-IL17F Rabbit Polyclonal Antibody

Supplier: BioVision
5109-50
10007-608EA 303.55 USD
10007-608
Anti-IL17F Rabbit Polyclonal Antibody
IL-17F, a member of the IL-17 family of structurally related cytokines, has been shown to stimulate proliferation and activation of T-cells and PBMCs. IL-17F also regulates cartilage matrix turnover and inhibits angiogenesis.

Recommended Dilutions: 1) Western Blot: Use 0.1-0.2 µg/ml. The detection limit for recombinant human IL-17F is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.<br>2) ELISA: Use 0.5 - 2.0 ?g/ml (100 ?l/well antibody solution) <br>3) Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of hIL-17F (50.0 ng/ml; a concentration of 0.9-1.2 ?g/ml of this antibody is required.

Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR