Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL17A Rabbit Polyclonal Antibody

Supplier: BioVision

5108-50
10007-606EA 252.35 USD InStock
10007-606
Anti-IL17A Rabbit Polyclonal Antibody
Antibodies
The originally described IL-17 protein, now known as IL-17A, is a homodimer of two 136 amino acid chains, secreted by activated T-cells that act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells.

Recommended Dilutions: 1) Western Blot: Use 0.1-0.2 µg/ml. The detection limit for recombinant human IL-17A is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.\

Type: Primary
Antigen: IL17A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty