Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GMCSF1 Rabbit Polyclonal Antibody

Supplier: BioVision

5100-100
10007-500EA 252.35 USD InStock
10007-500
Anti-GMCSF1 Rabbit Polyclonal Antibody
Antibodies
Human GM-CSF (Granulocyte Macrophage Colony Stimulating Factor) is a potent species-specific stimulator of bone marrow cells. GM-CSF stimulates precursor cells of granulocytes, macrophages and eosinophils.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty