Anti-Fractalkine Rabbit Polyclonal Antibody

Supplier: BioVision
5088-100
10007-490EA 286.04 USD
10007-490
Anti-Fractalkine Rabbit Polyclonal Antibody
Fractalkine, also known as neurotactin, is a membrane-bound CX3C Chemokine. The mature protein is part of a 397-amino acid precursor and consists of a chemokine domain of 76 amino acids, a mucin stalk of 241 amino acids, a putative transmembrane domain of 18 amino acids, and an intracellular tail of 37 amino acids. Within the chemokine domain, the first two cysteine residues are separated by 3 amino acids (CX3C). Fractalkine message is found in high abundance in the brain, kidney, lung and heart tissues. Fractalkine is chemotactic for monocytes and other leukocytes including NK cells and may play a role in brain inflammation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Fractalkine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR