Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Flt3 Ligand Rabbit Polyclonal Antibody

Supplier: BioVision

5085R-100
10007-488EA 252.35 USD InStock
10007-488
Anti-Flt3 Ligand Rabbit Polyclonal Antibody
Antibodies
Human flt3-Ligand (flk2-ligand) stimulates the proliferation and colony formation of certain bone marrow precursor cells including CD34+ cells. The antibody was produced using the recombinant human flt3-ligand as immunogen, which is a soluble 17.6 kDa polypeptide containing 155 amino acid residues, that comprises the extracellular domain of the transmembrane flt3-ligand protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Flt3 Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty