Anti-FGF-18 Rabbit Polyclonal Antibody

Supplier: BioVision
5082-100
10007-482EA 286.04 USD
10007-482
Anti-FGF-18 Rabbit Polyclonal Antibody
The FGFs plays an important role in regulating tissue growth, morphogenesis and tissue repair. FGF-18 is a newly discovered heparin binding growth factor that stimulates the proliferation and activation of cells that express FGF receptors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FGF-18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR