Anti-LAMP1 Rabbit Polyclonal Antibody

Supplier: BioVision
5077-100
10007-474EA 379.44 USD
10007-474
Anti-LAMP1 Rabbit Polyclonal Antibody
Autophagy, the process of bulk degradation of cellular proteins throμgh an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components and is negatively regulated by TOR (Target of rapamycin). A protein recently found to be involved in autophagy, LAMP-2, is a highly glycosylated protein associated with the lysosome. LAMP-1 shares much homology to LAMP-2 and is thoμght to have overlapping functions. Mice lacking LAMP-1 had very minor defects compared to those deficient in LAMP-2 expression. However, the loss of both proteins resulted in embryonic lethality, sμggesting that each protein possesses some unique and necessary functions.

Recommended Dilutions: Western Blot: 1 - 2 ?g/ml, Immunofluorescence: 20 ?g/ml, ELISA

Type: Primary
Antigen: LAMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR