Anti-FGF7 Rabbit Polyclonal Antibody

Supplier: BioVision
5050-100
10007-458EA 285.95 USD
10007-458
Anti-FGF7 Rabbit Polyclonal Antibody
FGF-7, also known as Keratinocyte Growth Factor (KGF), is a potent mitogen for keratinocytes and epithelial cells. The antibody recognizes ~19 kDa of human FGF-7/KGF protein containing 163 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: KGF/FGF-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR