Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Eotaxin/CCL11 Rabbit Polyclonal Antibody

Supplier: BioVision

5029-100
10007-434EA 272.95 USD InStock
10007-434
Anti-Eotaxin/CCL11 Rabbit Polyclonal Antibody
Antibodies
Eotaxin is a 74-amino acid eosinophil-chemotactic CC chemokine originally found in bronchoalveolar lavage fluid from allergic inflammatory subjects. It is involved in regulating the recruitment and activation of inflammatory leukocytes, particularly eosinophils. It may play a fundamental role in the development of allergic responses.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Eotaxin/CCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty