Anti-EGF Rabbit Polyclonal Antibody

Supplier: BioVision
5022-100
10007-430EA 286.04 USD
10007-430
Anti-EGF Rabbit Polyclonal Antibody
EGF (Epidermal Growth Factor) is polypeptide growth factor that stimulates the proliferation of a wide range of epidermal and epithelial cells. Human EGF is a 6.2 kDa protein containing 53 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR