Anti-PEG Rat Monoclonal Antibody [clone: 26A04]

Supplier: BioVision
5019-100
10007-428EA 286.04 USD
10007-428
Anti-PEG Rat Monoclonal Antibody [clone: 26A04]
Polyethylene Glycol (PEG) is a polymer of ethylene oxide available in size variants from 400 Da to 40 kDa. It is nonionic, nontoxic, biocompatible, strong, and hydrophilic and has a large exclusion volume in aqueous solution. PEG has a number of industrial and biomedical applications. The modification of a biopharmaceutical with PEG increases its hydrodynamic radius, reduces immunogenicity and proteolytic cleavage. In particular, therapeutic proteins are conjμgated with PEG to slow down their clearance from circulation and improve bioavailability. PEG antibodies can be a vital tool for propelling therapeutics to market by serving as a positive control anti-drμg antibody, measuring clearance of a drμg, or simply as a QA release confirming PEGylation.

Recommended Dilutions: N/A

Type: Primary
Antigen: PEG
Clonality: Monoclonal
Clone: 26A04
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR