Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PEG Mouse Monoclonal Antibody [clone: 09F02]

Supplier: BioVision

5018-100
10007-426EA 252.35 USD InStock
10007-426
Anti-PEG Mouse Monoclonal Antibody [clone: 09F02]
Antibodies
Polyethylene Glycol (PEG) is a polymer of ethylene oxide available in size variants from 400 Da to 40 kDa. It is nonionic, nontoxic, biocompatible, strong, hydrophilic and has a large exclusion volume in aqueous solution. PEG has a number of industrial and biomedical applications. The modification of a biopharmaceutical with PEG increases its hydrodynamic radius, reduces immunogenicity and proteolytic cleavage. In particular, therapeutic proteins are conjμgated with PEG to slow down their clearance from circulation and improve bioavailability. PEG antibodies can be a vital tool for propelling therapeutics to market by serving as a positive control anti-drμg antibody, measuring clearance of a drμg, or simply as a QA release confirming PEGylation.

Recommended Dilutions: N/A

Type: Primary
Antigen: PEG
Clonality: Monoclonal
Clone: 09F02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty