Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNTF Rabbit Polyclonal Antibody

Supplier: BioVision

5017-100
10007-424EA 252.35 USD InStock
10007-424
Anti-CNTF Rabbit Polyclonal Antibody
Antibodies
CNTF (Ciliary Neurotrophic Factor) is a potent neural factor that was originally characterized as a survivability factor for chick ciliary neurons in vitro. CNTF has also been shown to promote survivability and differentiation of other neuronal cell types. CNTF is a 22.9 kDa protein containing 200 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty