Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40L Rabbit Polyclonal Antibody

Supplier: BioVision

5015-100
10007-422EA 252.35 USD InStock
10007-422
Anti-CD40L Rabbit Polyclonal Antibody
Antibodies
Increasing evidence sμggests a central role for the CD40L-CD40 signaling pathway in several immunogenic and inflammatory processes, including atherosclerosis. It was also found that CD40L-CD40 signaling pathways play an important role in several autoimmune diseases, including the X-linked hyper-IgM syndrome, collagen-induced arthritis, allergic encephalitis and multiple sclerosis, and acute and chronic graft vs. host diseases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CD40L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty