Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Sushi Rabbit Polyclonal Antibody

Supplier: BioVision

5010-100
10007-414EA 334.75 USD InStock
10007-414
Anti-Sushi Rabbit Polyclonal Antibody
Antibodies
Sushi Peptide S3 is a trimer of one of the high endotoxin-binding domains, Sushi 3 (or S3) within Factor C, a lipopolysaccharide (LPS)-sensitive serine protease of the horseshoe crab (Limulus Polyphemus). S3 display detergent-like properties in disrupting LPS aggregates, with specificity for palmitoyl-oleoyl-phosphatidylglycerol (POPG) resulting from electrostatic and hydrophobic forces between the peptides and the bacterial lipids. The unsaturated nature of POPG confers fluidity and enhances insertion of the peptides into the lipid bilayer, causing maximal disruption of the bacterial membrane. In short, peptide S3 can bind to lipopolysaccharide (LPS) and inhibit the growth of Gram-negative bacteria without affecting mammalian cells. It has been shown that endotoxin activates Factor C based catalytic coagulation cascade resulting in the gelation of Limulus blood. This process is the basis of Limulus Amebocyte Lysate (LAL) endotoxin detection method.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: Sushi
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty