Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Amyloid beta 1-17 Mouse Monoclonal Antibody [clone: C4D10]

Supplier: BioVision

5006-100
10007-412EA 288.4 USD InStock
10007-412
Anti-Amyloid beta 1-17 Mouse Monoclonal Antibody [clone: C4D10]
Antibodies
Amyloid Beta (Aβ or Abeta) is a multi-functional peptide of 36-43 amino acids, processed from the Amyloid Precursor Protein (APP). It is most commonly known to be associated with Alzheimer's disease since it is the main component of amyloid plaques found in the brains of the diseased subjects. However, it also plays a role in multiple non-disease activities including protection against metal-induced reactive oxidizing species (ROS), modification of cholesterol transport, and potential activity as a transcription factor. It is secreted by platelets into the blood serum and its levels elevate with age.

Recommended Dilutions: Immunohistochemistry: 40-150 µg. However, optimal conditions must be determoned by the user.

Type: Primary
Antigen: Amyloid beta 1-17
Clonality: Monoclonal
Clone: C4D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human. The 1-13 Amino Acid Sequence Of This Protein Is Conserved In Mice, Rat, Humans And Other Species, So It Might Cross-Interact With Other Species’ Samples.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty