Anti-BDNF Rabbit Polyclonal Antibody

Supplier: BioVision
5004-100
10007-408EA 297.72 USD
10007-408
Anti-BDNF Rabbit Polyclonal Antibody
Human Brain-Derived Neurotrophic Factor (BDGF) is a potent neurotrophic factor that supports the growth and survivability of nerve and glial cells. BDNF protein is a 27.0 kDa dimer, formed by two identical 119 amino acid subunits held together by strong hydrophobic interactions. The antibody recognizes the mature form of BDNF at 14 kDa.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR