Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-6xHis Rabbit Polyclonal Antibody

Supplier: BioVision

3998-100
10005-202EA 288.4 USD InStock
10005-202
Anti-6xHis Rabbit Polyclonal Antibody
Antibodies
A variety of plasmids contain DNA that encodes an N-terminal tag consisting of six histidine (His) residues, followed by an extended multiple cloning sites. The 6xHis tag on the expressed recombinant proteins allows for efficient coupling to Ni++ affinity resins and purification by a single step chromatography. As is the case with other protein tags, this polyhistidine tag can be cleaved at sites recognized by enzymes such as thrombin and enterokinases to isolate the protein of interest.

Recommended Dilutions: Western Blot: 1 µg/ml; Immunoprecipitation: 40 µg/ml; and Immunocytochemistry: 2 µg/ml

Type: Primary
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty